missing translation for 'onlineSavingsMsg'
Learn More

Glycolic acid, MedChemExpress

Product Code. 16483449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483449 Supplier Medchem Express Supplier No. HYW015967
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glycolic acid is an inhibitor of tyrosinase, suppressing melanin formation and lead to a lightening of skin colour.

Glycolic acid is an inhibitor of tyrosinase, suppressing melanin formation and lead to a lightening of skin colour.

Specifications

Chemical Name or Material Glycolic acid
CAS 79-14-1
Color White
Purity Grade Notes Research
Molecular Formula C2H4O3
Quantity 1 g
Solubility Information H2O : 100 mg/mL (1314.92 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (1314.92 mM)
SMILES O=C(O)CO
Molecular Weight (g/mol) 76.05
Formula Weight 76.05
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.