missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 72.0°C to 80.0°C |
Boiling Point | 100.0°C |
Color | White |
Assay Percent Range | 99% |
Linear Formula | HOCH2CO2H |
Beilstein | 03, 228 |
Merck Index | 15, 4534 |
Formula Weight | 76.04 |
Percent Purity | 99% |
Quantity | 25 g |
Show More |
Safety and Handling
Product Identifier
- Glycolic acid
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'