missing translation for 'onlineSavingsMsg'
Learn More

Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate

Product Code. 11357136
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357136 1 g 1g
11367136 5 g 5g
11377136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357136 Supplier Thermo Scientific Alfa Aesar Supplier No. L07099.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30934-97-5 | C4H10O3 | 106.12 g/mol

Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30934-97-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00051799
InChI Key NYPNCQTUZYWFGG-UHFFFAOYSA-N
Synonym glycolaldehyde dimethyl acetal, ethanol, 2,2-dimethoxy, unii-u5use33y47, 2,2-dimethoxy-ethanol, 2,2-dimethoxy ethanol, glycolaldehydedimethylacetal, acmc-20aksv, 2,2-dimethoxyethan-1-ol, hydroxyacetaldehyde dimethylacetal, 2-hydroxyacetaldehyde dimethyl acetal
PubChem CID 542381
IUPAC Name 2,2-dimethoxyethanol
SMILES COC(CO)OC

Specifications

Melting Point <-76°C
Density 1.05
Boiling Point 68°C (21 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.413
Quantity 1 g
Beilstein 1697583
Solubility Information Miscible with water.
Formula Weight 106.12
Percent Purity 98%
Chemical Name or Material Glycolaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.