missing translation for 'onlineSavingsMsg'
Learn More

Glycolaldehyde-2-13C (aqueous solution), TRC

Product Code. 30391972
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391972 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391972 Supplier Toronto Research Chemicals Supplier No. TRCG6411015MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycolaldehyde-2-13C
CAS 478529-67-8
Molecular Formula C13CH4O2
Quantity 5 mg
Synonym 2-Hydroxyacetaldehyde-2-13C, 2-Hydroxyethanal-2-13C, 2-Oxoethanol-2-13C, Hydroxyacetaldehyde-2-13C, Diose-2-13C, Glicolaldehido-2-13C, Glycolic Aldehyde-2-13C, Glycollaldehyde-2-13C, Hydroxyethanal-2-13C, Methylolformaldehyde-2-13C, Monomethylolformaldehyde-2-13C, NSC 67935-2-13C
InChI Formula InChI=1S/C2H4O2/c3-1-2-4/h1,4H,2H2/i2+1
SMILES O[13CH2]C=O
IUPAC Name 2-hydroxyacetaldehyde
Molecular Weight (g/mol) 61.04
Formula Weight 61.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.