missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycol sulfite, 98%, Thermo Scientific Chemicals
£23.90 - £64.60
Chemical Identifiers
CAS | 3741-38-6 |
---|---|
Molecular Formula | C2H4O3S |
Molecular Weight (g/mol) | 108.11 |
MDL Number | MFCD00005354 |
InChI Key | WDXYVJKNSMILOQ-UHFFFAOYSA-N |
Synonym | ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol |
PubChem CID | 77342 |
IUPAC Name | 1,3,2-dioxathiolane 2-oxide |
SMILES | O=S1OCCO1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3741-38-6 | |
108.11 | |
WDXYVJKNSMILOQ-UHFFFAOYSA-N | |
77342 | |
O=S1OCCO1 |
C2H4O3S | |
MFCD00005354 | |
ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol | |
1,3,2-dioxathiolane 2-oxide |
Specifications
3741-38-6 | |
1.4200g/mL | |
79°C | |
98% | |
1.4450 to 1.447 | |
1.42 | |
WDXYVJKNSMILOQ-UHFFFAOYSA-N | |
1,3,2-dioxathiolane 2-oxide | |
77342 | |
98% | |
Glycol sulfite, 98% |
Colorless | |
68°C (20.0 mmHg) | |
Authentic | |
C2H4O3S | |
MFCD00005354 | |
ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol | |
O=S1OCCO1 | |
108.11 | |
108.11 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 223-131-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only