missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycol Salicylate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Salicylic acid-(2-hydroxyethyl) ester |
| CAS | 87-28-5 |
| Molecular Formula | C9 H10 O4 |
| Quantity | 50 g |
| Synonym | Hydroxyethyl Salicylate, Benzoic acid, 2-hydroxy-, 2-hydroxyethyl ester, Salicylic acid, 2-hydroxyethyl ester (6CI,7CI,8CI), Ethylene glycol, monosalicylate (8CI), 2-Hydroxyethyl 2-hydroxybenzoate, 2-Hydroxyethyl salicylate, Espirosal, Ethylene glycol salicylate, GL 7, Glycol monosalicylate, Glycol salicylate, Glysal, Monoglycol salicylate, Norgesic, Rheumacyl, Saliment, Sarocol, Spirosal, β-Hydroxyethyl salicylate |
| InChI Formula | InChI=1S/C9H10O4/c10-5-6-13-9(12)7-3-1-2-4-8(7)11/h1-4,10-11H,5-6H2 |
| SMILES | OCCOC(=O)c1ccccc1O |
| IUPAC Name | 2-hydroxyethyl 2-hydroxybenzoate |
| Molecular Weight (g/mol) | 182.17 |
| Formula Weight | 182.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction