missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen phosphorylase, muscle form Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16689
This item is not returnable.
View return policy
Description
Glycogen phosphorylase, muscle form Polyclonal antibody specifically detects Glycogen phosphorylase, muscle form in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| Glycogen phosphorylase, muscle form | |
| Polyclonal | |
| Unconjugated | |
| PBS pH7, 20% Glycerol with 0.025% Proclin 300 | |
| EC 2.4.1.1, glycogen phosphorylase, muscle form, myophosphorylase, phosphorylase, glycogen, muscle, phosphorylase, glycogen; muscle | |
| Rabbit | |
| 97 kDa | |
| 0.1 mL | |
| GPCR | |
| 5837 | |
| Human, Mouse, Rat, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.93mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P11217 | |
| PYGM | |
| Recombinant protein encompassing a sequence within the center region of human PYGM. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected to cross react based on sequence identity: Xenopus laevis (84%), Xenopus tropicalis (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction