missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Glycogen phosphorylase, muscle form Monoclonal antibody specifically detects Glycogen phosphorylase, muscle form in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Glycogen phosphorylase, muscle form |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 2C4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005600 |
| Gene Alias | EC 2.4.1.1, glycogen phosphorylase, muscle form, myophosphorylase, phosphorylase, glycogen, muscle, phosphorylase, glycogen, muscle |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?