missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen Phosphorylase BB/GPBB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20017
This item is not returnable.
View return policy
Description
Glycogen Phosphorylase BB/GPBB Polyclonal specifically detects Glycogen Phosphorylase BB/GPBB in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Glycogen Phosphorylase BB/GPBB | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB | |
| Rabbit | |
| 97 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.57mg/mL | |
| Western Blot 1:500-1:3000 | |
| P11216 | |
| PYGB | |
| Recombinant protein encompassing a sequence within the N-terminus region of human GPBB. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5834 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction