missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glycogen Phosphorylase BB/GPBB Antibody (E9B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27945
This item is not returnable.
View return policy
Description
Glycogen Phosphorylase BB/GPBB Monoclonal antibody specifically detects Glycogen Phosphorylase BB/GPBB in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Glycogen Phosphorylase BB/GPBB | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) | |
| Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB | |
| Synthetic peptide derived from human Glycogen Phosphorylase BB/GPBB | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| E9B10 | |
| Western Blot 1:1000 - 1:5000, ELISA | |
| P11216 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5834 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction