missing translation for 'onlineSavingsMsg'
Learn More

Glycogen, Mussel, MedChemExpress

Product Code. 16471047
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471047 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471047 Supplier Medchem Express Supplier No. HY113511
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glycogen is a glycolytic intermediates and high-energy phosphates that can serve as a form of energy storage in humans, animals, fungi, and bacteria.

Glycogen is a glycolytic intermediates and high-energy phosphates that can serve as a form of energy storage in humans, animals, fungi, and bacteria.

Specifications

Chemical Name or Material Glycogen, Mussel
Physical Form Powder
CAS 9005-79-2
Color White
Quantity 50 mg
Purity Grade Notes Research
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 50 mg/mL (Need ultrasonic)
SMILES [Glycogen]
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.