missing translation for 'onlineSavingsMsg'
Learn More

Glycodeoxycholic Acid, TRC

Product Code. 30402879
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402879 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402879 Supplier Toronto Research Chemicals Supplier No. TRCG6414031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycodeoxycholic Acid
CAS 360-65-6
Molecular Formula C26 H43 N O5
Quantity 1 g
Synonym Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, Glycine, N-(3α,12α-dihydroxy-5β-cholan-24-oyl)- (8CI), Glycodeoxycholic acid (6CI,7CI), 5β-Cholan-24-amide, N-(carboxymethyl)-3α,12α-dihydroxy- (8CI), Cholane, glycine deriv., N-[(3α,5β,12α)-3,12-Dihydroxy-24-oxocholan-24-yl]glycine, 3α,12α-Dihydroxy-5β-cholanic acid-24-glycine, Deoxycholic acid glycine conjugate, Deoxycholylglycine, Deoxyglycocholic acid, Glycodesoxycholic acid, Glycyldeoxycholic acid
InChI Formula InChI=1S/C26H43NO5/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-/m1/s1
SMILES C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name 2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
Molecular Weight (g/mol) 449.62
Formula Weight 449.3141
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.