missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycodeoxycholic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Glycodeoxycholic Acid Sodium Salt |
| CAS | 16409-34-0 |
| Molecular Formula | C26 H42 N O5 . Na |
| Quantity | 250 mg |
| Synonym | Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, sodium salt (1:1), Glycine, N-(3α,12α-dihydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, monosodium salt (9CI), Glycodeoxycholic acid, sodium salt (6CI,7CI), Cholane, glycine deriv., Sodium deoxyglycolate, Sodium glycodeoxycholate, Sodium glycyldeoxycholate, Glycodeoxycholic acid sodium salt, N-[(3α,5β,12α)-3,12-Dihydroxy-24-oxocholan-24-yl]glycine monosodium salt |
| InChI Formula | InChI=1S/C26H43NO5.Na/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3;/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;/m1./s1 |
| SMILES | [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC Name | sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate |
| Molecular Weight (g/mol) | 471.61 |
| Formula Weight | 471.2961 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?