missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycochenodeoxycholic Acid Sodium, 95%, TRC
Specifications
Specifications
| Chemical Name or Material | Glycochenodeoxycholic Acid Sodium Salt |
| CAS | 16564-43-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C26 H42 N O5 . Na |
| Quantity | 500 mg |
| Synonym | N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium, N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Monosodium, NSC 681056, Sodium Chenodeoxycholylglycine, Sodium Glycochenodeoxycholate, Sodium Glycochenodesoxycholate |
| InChI Formula | InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;/m1./s1 |
| SMILES | [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate |
| Molecular Weight (g/mol) | 471.61 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?