missing translation for 'onlineSavingsMsg'
Learn More

Glycochenodeoxycholic Acid-3-O-Beta-glucuronide Disodium Salt, TRC

Product Code. 30390485
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390485 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390485 Supplier Toronto Research Chemicals Supplier No. TRCG6412752.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycochenodeoxycholic Acid-3-O-beta-glucuronide Disodium Salt
CAS 75672-22-9
Purity Grade Notes HPLC
Molecular Formula C32H49NNa2O11
Quantity 2.5 mg
Synonym Cholane, β-D-Glucopyranosiduronic Acid Derivative, (3α,5β,7α)-24-[(Carboxymethyl)amino]-7-hydroxy-24-oxocholan-3-yl, β-D-Glucopyranosiduronic Acid Disodium Salt
InChI Formula InChI=1S/C32H51NO11.2Na/c1-15(4-7-22(35)33-14-23(36)37)18-5-6-19-24-20(9-11-32(18,19)3)31(2)10-8-17(12-16(31)13-21(24)34)43-30-27(40)25(38)26(39)28(44-30)29(41)42;;/h15-21,24-28,30,34,38-40H,4-14H2,1-3H3,(H,33,35)(H,36,37)(H,41,42);;/q;2*+1/p-2/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26+,27-,28+,30-,31+,32-;;/m1../s1
SMILES O=C(NCC(O[Na])=O)CC[C@@H](C)[C@](CC1)([H])[C@]2(C)[C@]1([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O[C@H]5[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O[Na])=O)O5)CC[C@]4(C)[C@@]3([H])CC2
IUPAC Name disodium;(2S,3S,4S,5R,6R)-6-[[(3R,5R,7R,8R,9S,10S,13R,14S,17R)-17-[(2R)-5-(carboxylatomethylamino)-5-oxopentan-2-yl]-7-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol) 669.71
Formula Weight 669.3101
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.