missing translation for 'onlineSavingsMsg'
Learn More

Glycitin, MedChemExpress

Product Code. 16488818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488818 10 mg 10mg
16483826 25 mg 25mg
16454727 50 mg 50mg
16441268 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488818 Supplier Medchem Express Supplier No. HYN0012
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glycitin is a natural isoflavone isolated from legumes; promotes the proliferation of bone marrow stromal cells and osteoblasts and suppresses bone turnover.Glycitin is antibacterial, antiviral and estrogenic.

Glycitin is a natural isoflavone isolated from legumes

Specifications

Chemical Name or Material Glycitin
Physical Form Solid
CAS 40246-10-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H22O10
For Use With (Application) Cancer-programmed cell death
Synonym Glycitein 7-O-β-glucoside
Solubility Information DMSO : ≥ 100 mg/mL (224.01 mM)
SMILES O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=CC(OC=C(C3=CC=C(O)C=C3)C4=O)=C4C=C2OC)[C@@H]1O
Molecular Weight (g/mol) 446.4
Formula Weight 446.4
Percent Purity 99.84%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.