missing translation for 'onlineSavingsMsg'
Learn More

Glycine, U.S.P. - F.C.C., J.T. Baker™

Catalog Number p-5391634
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O
This item is not returnable. View return policy
56-40-6
C2H5NO2
75.07
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
NCC(O)=O
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 75.07
Grade FCC/USP
Quantity 500 g
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'