missing translation for 'onlineSavingsMsg'
Learn More

Glycine, TRC

Product Code. 30006363
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006363 10 g 10g
17999334 10 mg 10mg
17959624 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006363 Supplier Toronto Research Chemicals Supplier No. TRCG61599010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2 H5 N O2
Molecular Weight (g/mol) 75.07
Synonym Glycine, 2-Aminoacetic acid, 2-Aminoacetic acid, Aciport, Aminoacetic acid, Aminoethanoic acid, Glicoamin, Glycocoll, Glycolixir, Glycosthene, Gyn-Hydralin, H 1, NSC 25936, NSC 2916, NSC 54188, Padil
IUPAC Name 2-aminoacetic acid
SMILES NCC(=O)O

Specifications

InChI Formula InChI=1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)
Quantity 10 g
Formula Weight 75.032
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Glycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.