missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-butyl ester hydrochloride, 99%

Product Code. 11363956
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363956 5 g 5g
11353956 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363956 Supplier Thermo Scientific Alfa Aesar Supplier No. L17116.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol

Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

Solubility
Soluble in methanol and water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00058255
InChI Key OSWULUXZFOQIRU-UHFFFAOYSA-N
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
PubChem CID 2725036
IUPAC Name tert-butyl 2-aminoacetate hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CN

Specifications

Melting Point ∽140°C (decomposition)
Beilstein 3989048
Solubility Information Soluble in methanol and water
Quantity 5 g
Formula Weight 167.64
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material Glycine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.