missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine tert butyl ester hydrochloride, 99%

Product Code. 10646481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10646481 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10646481 Supplier Thermo Scientific Chemicals Supplier No. 297020025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27532-96-3
Melting Point 138°C to 142°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H14ClNO2
Linear Formula H2NCH2C(O)OC(CH3)3·HCl
MDL Number MFCD00058255
Quantity 2.5 g
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
InChI Key OSWULUXZFOQIRU-UHFFFAOYSA-N
SMILES Cl.CC(C)(C)OC(=O)CN
IUPAC Name tert-butyl 2-aminoacetate hydrochloride
Molecular Weight (g/mol) 167.63
PubChem CID 2725036
Formula Weight 167.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glycine tert butyl ester hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.