missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-butyl ester, 97%

Product Code. 11362957
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362957

Brand: Thermo Scientific Alfa Aesar L16258.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6456-74-2 | C6H13NO2 | 131.175 g/mol

Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

Solubility
Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6456-74-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00038194
InChI Key SJMDMGHPMLKLHQ-UHFFFAOYSA-N
Synonym tert-butyl glycinate, glycine tert-butyl ester, glycine, 1,1-dimethylethyl ester, tert-butyl aminoacetate, h-gly-otbu, t-butyl glycinate, glycin-tert-butyl ester, 1,1-dimethylethyl glycinate, glycine t-butyl ester, aminoacetic acid tert-butyl ester
PubChem CID 151417
IUPAC Name tert-butyl 2-aminoacetate
SMILES CC(C)(C)OC(=O)CN

Specifications

Boiling Point 29°C to 31°C (2mmHg)
Refractive Index 1.424
Beilstein 2039000
Solubility Information Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.
Density 0.96
Quantity 1 g
Formula Weight 131.18
Sensitivity Air sensitive
Percent Purity 97%
Chemical Name or Material Glycine tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.