missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Receptor alpha 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03685-20UL
This item is not returnable.
View return policy
Description
Glycine Receptor alpha 2 Polyclonal specifically detects Glycine Receptor alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Glycine Receptor alpha 2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2742 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| GLR, glycine receptor alpha 2 subunit, glycine receptor subunit alpha-2, Glycine Receptor, Alpha polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-420 of human Glycine Receptor alpha 2 (NP_002054.1). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction