missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Receptor Alpha 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26171
This item is not returnable.
View return policy
Description
Glycine Receptor Alpha 1 Polyclonal specifically detects Glycine Receptor Alpha 1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.
Specifications
| Glycine Receptor Alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| GLRA1 | |
| Peptide with sequence CLQAKDGISVKGAN corresponding to internal region according to NP_037265.1. | |
| 0.1 mg | |
| Diabetes Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2741 | |
| Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| GLRA 1, Glycine receptor 48 kDa subunit, Glycine receptor strychnine-binding subunit, glycine receptor subunit alpha-1, glycine receptor, alpha 1, glycine receptor, alpha 1 (startle disease/hyperekplexia), MGC138878, MGC138879, STHE | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The immunizing peptide represents part of the cytoplasmic domain. The antibody is not expected to cross-react with other glycine receptor alpha proteins. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction