missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine N-Methyltransferase/GNMT Antibody (OTI8A3), DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glycine N-Methyltransferase/GNMT Monoclonal antibody specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
Specifications
| Antigen | Glycine N-Methyltransferase/GNMT |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI8A3 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.1.1.20, glycine N-methyltransferase |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human Glycine N-Methyltransferase/GNMT (NP_061833) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?