missing translation for 'onlineSavingsMsg'
Learn More

Glycine N-Methyltransferase/GNMT Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30732575 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732575 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732575 Supplier Novus Biologicals Supplier No. AF6526A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Glycine N-Methyltransferase/GNMT Polyclonal antibody specifically detects Glycine N-Methyltransferase/GNMT in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycine N-Methyltransferase/GNMT
Applications Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias EC 2.1.1.20, glycine N-methyltransferase
Host Species Sheep
Immunogen E. coli-derived recombinant human Glycine N-methyltransferase/GNMT, Met1-Asp295, Accession # Q14749
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 27232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.