missing translation for 'onlineSavingsMsg'
Learn More

Glycine, MP Biomedicals™

Product Code. 11407680 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407680 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407680 Supplier MP Biomedicals Supplier No. 02100570.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be synthesized in the body by Serine Hydroxymethyltransferase. Glycine, MP Biomedicals is a nonessential, non-chiral amino acid that acts as an inhibitory neurotransmitter, particularly in brain stem and medulla. It is commonly used as a buffer compon

Glycine does not have a chiral center, so it is the only amino acid with no asymmetric carbon. Therefore, it does not have a D- or L- isomer designation. This material is synthetic. It is the smallest of the 20 natural amino acids. It acs as inhibitory neurotransmitter that is shown to block NMDA-stimulated dopamine release in fetal mesencephalic cell cultures. It is also reported to increase NMDA-receptor mediated synaptic potential in cortical neurons.

Glycine is commonly used as a component in:

  • Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis
  • Towbin's transfer buffer for Western blots
  • Buffers used in cryoenzymology, osmotic pressure maintenance in isoelectric focusing of erythrocytes and salting-in effect in protein chemistry
  • Buffer for coupled phosphatase-kinase reaction for end labelling of restriction fragments
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-40-6
Molecular Formula C2H5NO2
MDL Number MFCD00008131
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
Solubility Information Soluble in water (250mg/mL at 25°C; 672mg/mL at 100°C); slightly soluble in alcohol and ethanol (95%); almost insoluble in ether.
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES NCC(O)=O
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.07
Quantity 500 g
PubChem CID 750
ChEBI CHEBI:15428
Formula Weight 75.07
Percent Purity ≥98%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.