missing translation for 'onlineSavingsMsg'
Learn More

Glycine Methyl Ester Hydrochloride, TRC

Product Code. 30395257
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395257 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395257 Supplier Toronto Research Chemicals Supplier No. TRCG62523510G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine Methyl Ester Hydrochloride
CAS 5680-79-5
Molecular Formula C3H8ClNO2
Quantity 10 g
Synonym Aminoacetic Acid Methyl Ester Hydrochloride, Glycinate Methyl Ester Hydrochloride, Glycine Methyl Ester Monohydrochloride, Methyl 2-Aminoacetate Hydrochloride, Methyl Aminoacetate Hydrochloride, Methyl Glycinate Hydrochloride, Methyl alpha-Aminoacetate Hydrochloride
InChI Formula InChI=1S/C3H7NO2.ClH/c1-6-3(5)2-4;/h2,4H2,1H3;1H
SMILES Cl.COC(=O)CN
IUPAC Name methyl 2-aminoacetate;hydrochloride
Molecular Weight (g/mol) 125.55
Formula Weight 125.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.