missing translation for 'onlineSavingsMsg'
Learn More

Glycine methyl ester hydrochloride, 99%

Product Code. 11458026
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458026 2500 g 2500g
11438026 100 g 100g
11448026 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458026 Supplier Thermo Scientific Alfa Aesar Supplier No. A14083.0E
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol

Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

Solubility
Soluble in water (2% w/v) and methanol (50 mg/ml).

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-79-5
Molecular Formula C3H8ClNO2
Molecular Weight (g/mol) 125.55
MDL Number MFCD00012870
InChI Key COQRGFWWJBEXRC-UHFFFAOYSA-N
Synonym glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
PubChem CID 122755
IUPAC Name methyl 2-aminoacetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CN

Specifications

Melting Point ∼175°C (decomposition)
Beilstein 3593644
Solubility Information Soluble in water (2% w/v) and methanol (50mg/ml).
Density 1
Quantity 2500 g
Formula Weight 125.56
Percent Purity 99%
Chemical Name or Material Glycine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.