missing translation for 'onlineSavingsMsg'
Learn More

Glycine methyl ester hydrochloride, 98%

Product Code. 10255250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255250 100 g Plastic bottle 100g
10510931 500 g Plastic bottle 500g
13498318 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255250 Supplier Thermo Scientific Acros Supplier No. 120101000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-79-5
Molecular Formula C3H8ClNO2
Molecular Weight (g/mol) 125.55
MDL Number MFCD00012870
InChI Key COQRGFWWJBEXRC-UHFFFAOYSA-N
Synonym glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
PubChem CID 122755
IUPAC Name methyl 2-aminoacetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CN

Specifications

Melting Point 173°C to 177°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula H2NCH2CO2CH3·HCl
Quantity 100 g
Beilstein 04,34
Solubility Information Solubility in water: >1000g/L water (20 c)
Formula Weight 125.56
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Glycine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.