missing translation for 'onlineSavingsMsg'
Learn More

Glycine hydrochloride, 98%

Product Code. 11404793
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404793 2500 g 2500g
11484783 100 g 100g
11494783 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404793 Supplier Thermo Scientific Alfa Aesar Supplier No. A12162.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6000-43-7 | C2H6ClNO2 | 111.53 g/mol

Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6000-43-7
Molecular Formula C2H6ClNO2
Molecular Weight (g/mol) 111.53
MDL Number MFCD00012872
InChI Key IVLXQGJVBGMLRR-UHFFFAOYSA-N
Synonym glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
PubChem CID 22316
IUPAC Name 2-aminoacetic acid;hydrochloride
SMILES [H+].[Cl-].NCC(O)=O

Specifications

Melting Point 175°C to 178°C
Beilstein 3909617
Merck Index 14,4491
Solubility Information Soluble in water (>100g/L).
Quantity 2500 g
Odor Odorless
Formula Weight 111.53
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Glycine hydrochloride
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.