missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Glycine Free Base
Description
For use in preparing electrophoresis buffers.
For use in preparing electrophoresis buffers. Cell culture tested.
Specifications
Specifications
| Chemical Name or Material | Glycine, Free Base |
| CAS | 56-40-6 |
| Molecular Formula | C2H5NO2 |
| Quantity | 5 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?