missing translation for 'onlineSavingsMsg'
Learn More

Glycine ethyl ester hydrochloride, 99%

Product Code. 11404083
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11404083

Brand: Thermo Scientific Alfa Aesar A10315.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-33-6 | C4H10ClNO2 | 139.579 g/mol

Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N?-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

Solubility
Soluble in water (>1000 mg/ml at 20°C), ethanol, DMSO, and methanol.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-33-6
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.579
MDL Number MFCD00012871
InChI Key TXTWXQXDMWILOF-UHFFFAOYSA-N
Synonym glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
PubChem CID 2723640
IUPAC Name ethyl 2-aminoacetate;hydrochloride
SMILES CCOC(=O)CN.Cl

Specifications

Melting Point 144°C to 148°C
Beilstein 3594034
Solubility Information Soluble in water (>1000mg/ml at 20°C),ethanol,DMSO,and methanol.
Quantity 2500 g
Formula Weight 139.58
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material Glycine ethyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.