missing translation for 'onlineSavingsMsg'
Learn More

Glycine ethyl ester hydrochloride, 99%

Product Code. 10275200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275200 500 g Plastic bottle 500g
10326610 100 g Plastic bottle 100g
11965331 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275200 Supplier Thermo Scientific Acros Supplier No. 120095000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-33-6 | C4H9NO2·HCl | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-33-6
Molecular Formula C4H9NO2·HCl
Molecular Weight (g/mol) 139.58
InChI Key TXTWXQXDMWILOF-UHFFFAOYSA-N
Synonym glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
PubChem CID 2723640
IUPAC Name ethyl 2-aminoacetate;hydrochloride
SMILES CCOC(=O)CN.Cl

Specifications

Melting Point 145°C to 148°C
Color White
pH ≥3 (1% aq. soln.)
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CO2C2H5·HCl
Quantity 500 g
Beilstein 04,34
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in ethanol
Formula Weight 139.58
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glycine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.