missing translation for 'onlineSavingsMsg'
Learn More

Glycine ethyl ester hydrochloride, 99%

Product Code. 11494073
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494073 500 g 500g
11484073 100 g 100g
11404083 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494073 Supplier Thermo Scientific Alfa Aesar Supplier No. A10315.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-33-6 | C4H10ClNO2 | 139.579 g/mol

Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N?-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

Solubility
Soluble in water (>1000 mg/ml at 20°C), ethanol, DMSO, and methanol.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-33-6
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.579
MDL Number MFCD00012871
InChI Key TXTWXQXDMWILOF-UHFFFAOYSA-N
Synonym glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
PubChem CID 2723640
IUPAC Name ethyl 2-aminoacetate;hydrochloride
SMILES CCOC(=O)CN.Cl

Specifications

Melting Point 144°C to 148°C
Beilstein 3594034
Solubility Information Soluble in water (>1000mg/ml at 20°C),ethanol,DMSO,and methanol.
Quantity 500 g
Formula Weight 139.58
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material Glycine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.