missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Dr. Ehrenstorfer

Product Code. 30321581
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321581 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321581 Supplier Dr. Ehrenstorfer Supplier No. DREC14037000100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine
CAS 56-40-6
Analyte or Component Names Glycine
Shipping Condition +20°C
Molecular Formula C2 H5 N O2
Certifications/Compliance ISO 17034
Synonym Glycine, 2-Aminoacetic acid, 2-Aminoacetic acid, 20: PN: WO2012076822 PAGE: 51 claimed sequence, 5: PN: US20090069547 PAGE: 10 claimed protein, 5: PN: WO2013027191 SEQID: 13 claimed sequence, 68: PN: US20110195077 SEQID: 13 claimed sequence, 75: PN: WO2011084714 SEQID: 78 claimed sequence, Aciport, Aminoacetic acid, Aminoethanoic acid, Glicoamin, Glycocoll, Glycolixir, Glycosthene, Gyn-Hydralin, H 1, NSC 25936, NSC 2916, NSC 54188, Padil
InChI Formula InChI=1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)
SMILES NCC(=O)O
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.07
Quantity 100 mg
Formula Weight 75.032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.