missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Crystalline MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
Specifications
Specifications
| Density | 1.16 g/mL |
| Quantity | 1 kg |
| Formula Weight | 75.07 |
| Physical Form | Crystalline |
| Chemical Name or Material | Glycine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction