missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Buffer Component for Electrophoresis and Blotting, Fisher Chemical

Product Code. 10080160 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10080160 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10080160 Supplier Fisher Chemical Supplier No. G/P460/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 182°C
CAS 56-40-6
pH 5.9 to 6.4
Color White
Molecular Formula C2H5NO2
Packaging Plastic Powder Jar
MDL Number MFCD00008131
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES NCC(O)=O
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.07
Quantity 500 g
PubChem CID 750
ChEBI CHEBI:15428
Formula Weight 75.07g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.