missing translation for 'onlineSavingsMsg'
Learn More

Glycine Benzyl Ester p-Toluenesulfonate, TRC

Product Code. 30402336
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402336 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402336 Supplier Toronto Research Chemicals Supplier No. TRCG625100100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine Benzyl Ester, p-Toluenesulfonate
CAS 1738-76-7
Molecular Formula C9 H11 N O2 . C7 H8 O3 S
Quantity 100 g
Synonym Glycine Phenylmethyl Ester 4-Methylbenzenesulfonate (1:1), Glycine, benzyl ester, p-toluenesulfonate (6CI,8CI), Glycine, phenylmethyl ester, 4-methylbenzenesulfonate (9CI), Benzyl glycinate p-toluenesulfonate, Benzyl glycinate p-toluenesulfonic acid salt, Benzyl glycinate p-tolylsulfonate, Benzyl glycinate toluenesulfonate, Benzyl glycinate tosylate, Gly-OBzl TosOH, Glycine benzyl ester 4-toluenesulfonate, Glycine benzyl ester 4-toluenesulfonic acid salt, Glycine benzyl ester p-tosylate, Glycine benzyl ester tosylate, H-Gly-OBzl toluenesulfonate
InChI Formula InChI=1S/C9H11NO2.C7H8O3S/c10-6-9(11)12-7-8-4-2-1-3-5-8;1-6-2-4-7(5-3-6)11(8,9)10/h1-5H,6-7,10H2;2-5H,1H3,(H,8,9,10)
SMILES Cc1ccc(cc1)S(=O)(=O)O.NCC(=O)OCc2ccccc2
IUPAC Name benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 337.39
Formula Weight 337.0984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.