missing translation for 'onlineSavingsMsg'
Learn More

Glycine benzyl ester p-toluenesulfonate salt, 99%

Product Code. 15462438
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462438 5 g 5g
15422416 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462438 Supplier Thermo Scientific Alfa Aesar Supplier No. H56016.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1738-76-7 | C16H19NO5S | 337.39 g/mol

Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.

Solubility
Soluble in methanol. (0.1 g/mL)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-76-7
Molecular Formula C16H19NO5S
Molecular Weight (g/mol) 337.39
MDL Number MFCD00035425
InChI Key WJKJXKRHMUXQSL-UHFFFAOYSA-N
Synonym h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
PubChem CID 6451311
IUPAC Name benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN

Specifications

Melting Point 132°C to 134°C
Linear Formula NH2CH2COOCH2C6H5·CH3C6H4SO3H
Quantity 5 g
Beilstein 3600116
Solubility Information Soluble in methanol. (0.1g/mL)
Formula Weight 337.39
Percent Purity 99%
Chemical Name or Material Glycine benzyl ester p-toluenesulfonate salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.