missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, ACS, 98.5+%

Product Code. 11335837
Click to view available options
Quantity:
100 g
500 g
3 kg
Unit Size:
100g
3kg
500g
This item is not returnable. View return policy

Product Code. 11335837

Brand: Thermo Scientific Chemicals 043497.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

Glycine is employed to protect the kidneys from the harmful side effects of certain drugs used after organ transplantation as well as the liver from the harmful effects of alcohol. The solution of glycine plays an important role for deposition of alloys and metals. It is likewise useful for patients with acute alcohol hallucinosis. It is applied for treating schizophrenia, stroke, benign prostatic hyperplasia (BPH), and some rare inherited metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
pH 5.97
Color White
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in pyridine,water. Sparingly soluble in ethanol. Insoluble in ether.
Density 1.595 g/mL
Quantity 500 g
Odor Odorless
Formula Weight 75.07
Percent Purity ≥98.5%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.