missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals CSM-URA w/40 mg/L ADE (Powder)
Shop All MP Biomedicals Products
Click to view available options
:
100 g
Description
- Single dropout formulation (without uracil) with elevated adenine.
- The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine).
- Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component.
- This CSM-Dropout medium creates a richer medium for better yield and growth rate, and increases the probability of successful transformations when screening libraries or performing gene knock-outs.
- Research Use Only (RUO).
Specifications
Specifications
| Appearance | Clear to Slightly Hazy |
| Certifications/Compliance | Molecular Biology Certified™ |
| Color | Colorless to faint yellow |
| Content And Storage | Store dry powder at ambient temperature (15-30°C). Powder medium is hygroscopic; tightly cap; minimize exposure to air. Store reconstituted media at 4°C. |
| Form | Powder |
| Grade | Molecular Biology Certified™ |
| Media Type | Yeast media |
| Formulation | Without uracil |
| For Use With (Application) | Cell culture, Proteomics, Purification and Detection, Protein Analysis, Yeast Culture, Plasmid Production, Protein Expression, Gene Expression, Cloning, Gene Function, Gene Regulation, Automated Bacterial Growth Media, S. cerevisiae Culture |
| No. per Pack | 100 g/Pk. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction