missing translation for 'onlineSavingsMsg'
Learn More

Glycine, MP Biomedicals™

Product Code. 11490612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
3 kg
Unit Size:
250g
3kg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490612 50 g 50g
11400622 250 g 250g
11488272 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490612 Supplier MP Biomedicals Supplier No. 0219404950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine, ACS Reagent Grade, White Crystalline Powder

  • It is widely used as a buffer for a variety of immunological applications.
  • It is also used as an eluent in column chromatography.
  • It has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
  • Commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots , a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
  • Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon.
  • It is a major inhibitory neurotransmitter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 240°C (Decomposition)(Literature)
Color White
Assay Percent Range ≥98.5%
Beilstein 635782
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
Solubility Information Soluble in water (250mg/mL at 25°C; 672mg/mL at 100°C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
Density 1.16g/mL (Lit.)
Quantity 50 g
Formula Weight 75.07
Percent Purity ≥98.5%
Grade Reagent
Physical Form Crystalline Powder
Assay ≥98.5%
Chemical Name or Material Glycine, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.