missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine-2-13C Ethyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Glycine-2-13C Ethyl Ester, Hydrochloride |
| CAS | 58420-91-0 |
| Molecular Formula | 13C C3 H9 N O2 . H Cl |
| Quantity | 250 mg |
| Synonym | Glycine-2-13C, ethyl ester, hydrochloride (1:1), Glycine-2-13C, ethyl ester, hydrochloride (9CI), Glycine-2-13C ethyl ester hydrochloride, Glycine-13C ethyl ester hydrochloride, 2-Aminoacetic-2-13C acid ethyl ester hydrochloride, Ethyl 2-aminoacetate-2-13C hydrochloride, Ethyl glycinate-2-13C hydrochloride, Ethyl glycine-13C hydrochloride |
| InChI Formula | InChI=1S/C4H9NO2.ClH/c1-2-7-4(6)3-5;/h2-3,5H2,1H3;1H/i3+1; |
| SMILES | Cl.CCOC(=O)[13CH2]N |
| IUPAC Name | ethyl 2-aminoacetate;hydrochloride |
| Molecular Weight (g/mol) | 140.57 |
| Formula Weight | 140.0434 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?