missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine-15N,d2, TRC
Specifications
Specifications
| Chemical Name or Material | Glycine-15N,d2 |
| Molecular Formula | C2 D2 H3 15N O2 |
| Quantity | 100 mg |
| Synonym | 2-Aminoacetic Acid-15N,d2, Aciport-15N,d2, Aminoethanoic Acid-15N,d2, Glicoamin-15N,d2, Glycocoll-15N,d2, Glycolixir-15N,d2, Glycosthene-15N,d2, Gyn-Hydralin-15N,d2, H 1-15N,d2, NSC 25936-15N,d2, NSC 2916-15N,d2, NSC 54188-15N,d2, Padil-15N,d2 |
| InChI Formula | InChI=1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1D2,3+1 |
| SMILES | [2H]C([2H])([15NH2])C(=O)O |
| IUPAC Name | 2-(15N)azanyl-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 78.072 |
| Formula Weight | 78.042 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?