missing translation for 'onlineSavingsMsg'
Learn More

Glycine-13D2,15N Methyl Ester Hydrochloride, TRC

Product Code. 30400433
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400433 2.5 mg 2.5mg
30408659 5 mg 5mg
30392833 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400433 Supplier Toronto Research Chemicals Supplier No. TRCG6160122.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine-13C2,15N Methyl Ester Hydrochloride
Molecular Formula C13C2H715NO2.HCl
Quantity 2.5 mg
Synonym Methyl 2-Aminoacetate-13C2,15N Hydrochloride
InChI Formula InChI=1S/C3H7NO2.ClH/c1-6-3(5)2-4;/h2,4H2,1H3;1H/i2+1,3+1,4+1;
SMILES [15NH2][13CH2][13C](OC)=O.Cl
IUPAC Name methyl glycinate-13C2-15N hydrochloride
Molecular Weight (g/mol) 128.53
Formula Weight 128.03
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.