missing translation for 'onlineSavingsMsg'
Learn More

Glycine-13C2 Ethyl Ester Hydrochloride, TRC

Product Code. 30400148
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400148 Supplier Toronto Research Chemicals Supplier No. TRCG625228100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine-13C2 Ethyl Ester Hydrochloride
CAS 1246819-31-7
Molecular Formula 13C2 C2 H9 N O2 . H Cl
Quantity 100 mg
Synonym Glycine-1,2-13C2, ethyl ester, hydrochloride (1:1), Glycine-13C2 ethyl ester hydrochloride, 2-Aminoacetic-1,2-13C2 acid ethyl ester hydrochloride, Ethyl 2-aminoacetate-1,2-13C2 hydrochloride, Ethyl glycinate-13C2 hydrochloride, Ethyl glycine-13C2 hydrochloride
InChI Formula InChI=1S/C4H9NO2.ClH/c1-2-7-4(6)3-5;/h2-3,5H2,1H3;1H/i3+1,4+1;
SMILES Cl.CCO[13C](=O)[13CH2]N
IUPAC Name ethyl 2-aminoacetate;hydrochloride
Molecular Weight (g/mol) 141.57
Formula Weight 141.0467
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.