missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine-13C2 Ethyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Glycine-13C2 Ethyl Ester Hydrochloride |
| CAS | 1246819-31-7 |
| Molecular Formula | 13C2 C2 H9 N O2 . H Cl |
| Quantity | 10 mg |
| Synonym | Glycine-1,2-13C2, ethyl ester, hydrochloride (1:1), Glycine-13C2 ethyl ester hydrochloride, 2-Aminoacetic-1,2-13C2 acid ethyl ester hydrochloride, Ethyl 2-aminoacetate-1,2-13C2 hydrochloride, Ethyl glycinate-13C2 hydrochloride, Ethyl glycine-13C2 hydrochloride |
| InChI Formula | InChI=1S/C4H9NO2.ClH/c1-2-7-4(6)3-5;/h2-3,5H2,1H3;1H/i3+1,4+1; |
| SMILES | Cl.CCO[13C](=O)[13CH2]N |
| IUPAC Name | ethyl 2-aminoacetate;hydrochloride |
| Molecular Weight (g/mol) | 141.57 |
| Formula Weight | 141.0467 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?