missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.5

Product Code. p-7041762
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15482969

Brand: Thermo Scientific Chemicals J60154.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-40-675.067 g/mol

Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
It is soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Weight (g/mol) 75.067

Specifications

Concentration 0.2M buffer soln.
pH 3.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.