missing translation for 'onlineSavingsMsg'
Learn More

Glycinamide hydrochloride, 98%

Product Code. 11469023
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469023 25 g 25g
11459023 5 g 5g
11479023 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469023 Supplier Thermo Scientific Alfa Aesar Supplier No. A11128.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1668-10-6 | C2H7ClN2O | 110.541 g/mol

Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

Solubility
Soluble in water (1100g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1668-10-6
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00013008
InChI Key WKNMKGVLOWGGOU-UHFFFAOYSA-N
Synonym glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
PubChem CID 2723639
IUPAC Name 2-aminoacetamide;hydrochloride
SMILES C(C(=O)N)N.Cl

Specifications

Melting Point ∼204°C (decomposition)
Quantity 25 g
Beilstein 3554199
Sensitivity Hygroscopic
Solubility Information Soluble in water (1100g/L).
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material Glycinamide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.