missing translation for 'onlineSavingsMsg'
Learn More

Glycinal Dimethyl Acetal, TRC

Product Code. 30396002
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396002 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396002 Supplier Toronto Research Chemicals Supplier No. TRCG616495500ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycinal Dimethyl Acetal
CAS 22483-09-6
Molecular Formula C4H11NO2
Quantity 500 mL
Synonym Aminoacetaldehyde Dimethylacetal, 2,2-Dimethoxyethanamine, 2,2-Dimethoxyethylamine, 2-Aminoacetaldehyde Dimethyl Acetal, Aminoacetaldehyde Dimethyl Acetal, DEA, NSC 73701
InChI Formula InChI=1S/C4H11NO2/c1-6-4(3-5)7-2/h4H,3,5H2,1-2H3
SMILES COC(CN)OC
IUPAC Name 2,2-dimethoxyethanamine
Molecular Weight (g/mol) 105.14
Formula Weight 105.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.